Product Details

SNP ID
rs141600690
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:2529026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTTTCCTATGAGGATACAGCT[C/T]GAGTTCGGCCTTCACAAAAGGCAAA
Phenotype
MIM: 607348 MIM: 606162
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HES5 PubMed Links

Gene Details

Gene
HES5
Gene Name
hes family bHLH transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010926.3 1025 UTR 3 NP_001010926.1
XM_005244751.4 1025 UTR 3 XP_005244808.1
Gene
PANK4
Gene Name
pantothenate kinase 4
There are no transcripts associated with this gene.

View Full Product Details