Product Details

SNP ID
rs141824919
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:248626687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAAAGCCATCGAGGGAGCCCC[C/T]AAACCAGGCACCAGCAGCCAGCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2T10 PubMed Links

Gene Details

Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 442 Intron NP_001004693.1
XM_017000273.1 442 Intron XP_016855762.1
Gene
OR2T11
Gene Name
olfactory receptor family 2 subfamily T member 11 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001964.1 442 Missense Mutation AGG,GGG R148G NP_001001964.1

View Full Product Details