Product Details

SNP ID
rs141927084
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:37813673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGGGCTCTTGGGAGGCAGAGG[C/T]CTATTCTGCACCTCAGGCCATGCCT
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005955.2 4030 UTR 3 NP_005946.2
XM_011541491.2 4030 UTR 3 XP_011539793.1
XM_011541493.2 4030 Intron XP_011539795.1
XM_011541494.1 4030 UTR 3 XP_011539796.1
Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
There are no transcripts associated with this gene.

View Full Product Details