Product Details

SNP ID
rs142012185
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:24121004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGAGGTTGCAAAGGGAGGGAC[A/T]TGGGGTGGCCCTCGATCTGGACTGA
Phenotype
MIM: 605457 MIM: 616832
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL22RA1 PubMed Links

Gene Details

Gene
IL22RA1
Gene Name
interleukin 22 receptor subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021258.3 1729 Missense Mutation AAG,ATG K509M NP_067081.2
XM_011541882.1 1729 Missense Mutation AAG,ATG K373M XP_011540184.1
Gene
MYOM3
Gene Name
myomesin 3
There are no transcripts associated with this gene.

View Full Product Details