Product Details

SNP ID
rs142281141
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247100767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTATCACATTTGGTACATTT[A/G]TAGGGTCTTTCTCCAGTGTGAGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf229 PubMed Links

Gene Details

Gene
C1orf229
Gene Name
chromosome 1 open reading frame 229
There are no transcripts associated with this gene.

Gene
ZNF669
Gene Name
zinc finger protein 669
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142572.1 884 Silent Mutation TAC,TAT Y248Y NP_001136044.1
NM_024804.2 884 Silent Mutation TAC,TAT Y334Y NP_079080.2
XM_005273270.2 884 Silent Mutation TAC,TAT Y211Y XP_005273327.1
XM_005273271.2 884 Silent Mutation TAC,TAT Y211Y XP_005273328.1

View Full Product Details