Product Details

SNP ID
rs142293421
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:228174954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGGTGGCGGCTCCTCACCCAG[C/G]ATGAAGGCCCAGCCCTGGTGCCCGG
Phenotype
MIM: 615316
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IBA57 PubMed Links

Gene Details

Gene
IBA57
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010867.3 613 Missense Mutation CAT,GAT H202D NP_001010867.1
NM_001310327.1 613 Missense Mutation CAT,GAT H9D NP_001297256.1
Gene
IBA57-AS1
Gene Name
IBA57 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details