Product Details

SNP ID
rs142355297
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:108817143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGGGTGAGGGTGTTGCAGGGT[C/T]TGAGCTGTAGGTCATGAAAGCCTGA
Phenotype
MIM: 608339
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKNAD1 PubMed Links

Gene Details

Gene
AKNAD1
Gene Name
AKNA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152763.4 2563 Missense Mutation AAC,GAC N762D NP_689976.2
Gene
STXBP3
Gene Name
syntaxin binding protein 3
There are no transcripts associated with this gene.

View Full Product Details