Product Details

SNP ID
rs142370840
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153986227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACTTGAAGAGGTGGTCGTAGG[C/T]TTTGGCCATGGCGGACACCGGGGGA
Phenotype
MIM: 604671 MIM: 602672 MIM: 603702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JTB PubMed Links

Gene Details

Gene
JTB
Gene Name
jumping translocation breakpoint
There are no transcripts associated with this gene.

Gene
NUP210L
Gene Name
nucleoporin 210 like
There are no transcripts associated with this gene.

Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272038.1 151 Intron NP_001258967.1
NM_002870.3 151 Missense Mutation ACC,GCC T4A NP_002861.1
XM_017001959.1 151 Intron XP_016857448.1
Gene
RPS27
Gene Name
ribosomal protein S27
There are no transcripts associated with this gene.

View Full Product Details