Product Details

SNP ID
rs142663259
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:151762426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTCAGGGGATGCATATACAG[C/T]CAGTCCCTGAGGAAGGAGTCGATTC
Phenotype
MIM: 611824 MIM: 605138 MIM: 609501
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPL9 PubMed Links
Additional Information
For this assay, SNP(s) [rs56289123] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MRPL9
Gene Name
mitochondrial ribosomal protein L9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300733.1 822 Missense Mutation ACT,GCT T129A NP_001287662.1
NM_031420.3 822 Missense Mutation ACT,GCT T129A NP_113608.1
Gene
OAZ3
Gene Name
ornithine decarboxylase antizyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134939.1 822 Intron NP_001128411.1
NM_001301371.1 822 Intron NP_001288300.1
NM_016178.2 822 Intron NP_057262.2
Gene
TDRKH
Gene Name
tudor and KH domain containing
There are no transcripts associated with this gene.

View Full Product Details