Product Details

SNP ID
rs142759531
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:85582103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTGTCCCAACCCTCGGCTGGT[C/G]AAAGTTACCGGGCAGTGCTGCGAGG
Phenotype
MIM: 602369 MIM: 604743
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYR61 PubMed Links

Gene Details

Gene
CYR61
Gene Name
cysteine rich angiogenic inducer 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001554.4 677 Silent Mutation GTC,GTG V151V NP_001545.2
Gene
DDAH1
Gene Name
dimethylarginine dimethylaminohydrolase 1
There are no transcripts associated with this gene.

View Full Product Details