Product Details

SNP ID
rs142791991
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:89825757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTAGGAAATTACCATTCCCAGA[C/T]GTCTTCTGAAGCTGGCAGGTAAGAA
Phenotype
MIM: 612890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC8D PubMed Links

Gene Details

Gene
LRRC8D
Gene Name
leucine rich repeat containing 8 family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134479.1 Intron NP_001127951.1
NM_018103.4 Intron NP_060573.2
XM_011541689.1 Intron XP_011539991.1
XM_017001599.1 Intron XP_016857088.1
XM_017001600.1 Intron XP_016857089.1
XM_017001601.1 Intron XP_016857090.1
XM_017001602.1 Intron XP_016857091.1

View Full Product Details