Product Details

SNP ID
rs142809756
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:158547197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTATGGTCTTTCTGAGGGCTGC[C/G]TTTACTTCATGGTTCCTCAGACAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6Y1 PubMed Links

Gene Details

Gene
OR6Y1
Gene Name
olfactory receptor family 6 subfamily Y member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005189.1 909 Missense Mutation AAC,AAG N303K NP_001005189.1

View Full Product Details