Product Details

SNP ID
rs142836578
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:206897800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAAGAAGAATTGAGGCTGCTTG[A/G]GAGGAAGGCCAGGAGGAACACGAGA
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 243 UTR 5 NP_001172085.1
NM_001185157.1 243 UTR 5 NP_001172086.1
NM_001185158.1 243 UTR 5 NP_001172087.1
NM_006850.3 243 UTR 5 NP_006841.1
XM_011509101.2 243 Intron XP_011507403.1
XM_017000121.1 243 Intron XP_016855610.1

View Full Product Details