Product Details

SNP ID
rs142954972
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:10463134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTAAGGATGTGGCTCGCCAGC[A/G]CAACGTCCGAGGAGGTCTCTCTGCT
Phenotype
MIM: 601882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DFFA PubMed Links

Gene Details

Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004401.2 805 Missense Mutation GCG,GTG A236V NP_004392.1
NM_213566.1 805 Missense Mutation GCG,GTG A236V NP_998731.1

View Full Product Details