Product Details

SNP ID
rs143080465
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:156299054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCTCGCGTGAGTTCACAGAGC[A/G]CAGCACAGGCCATGCTGCTCTGGCT
Phenotype
MIM: 600114 MIM: 610962 MIM: 615531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
There are no transcripts associated with this gene.

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
There are no transcripts associated with this gene.

Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004319.2 584 Missense Mutation CGC,TGC R46C NP_001004319.1

View Full Product Details