Product Details

SNP ID
rs143117795
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:37613805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCATGTAGCACCCTGCGTGTC[C/T]GCTCCTCGGAGCCCCTCCGGAAACC
Phenotype
MIM: 609595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPO1 PubMed Links

Gene Details

Gene
RSPO1
Gene Name
R-spondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038633.3 1416 Missense Mutation CAG,CGG Q175R NP_001033722.1
NM_001242908.1 1416 Missense Mutation CAG,CGG Q175R NP_001229837.1
NM_001242909.1 1416 Missense Mutation CAG,CGG Q148R NP_001229838.1
NM_001242910.1 1416 Intron NP_001229839.1
XM_006710583.3 1416 Missense Mutation CAG,CGG Q175R XP_006710646.1

View Full Product Details