Product Details

SNP ID
rs143127732
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:114707764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTTCACGTTTGCGGTTTGGTT[C/T]TCTGTTTCAGACATGAACTGCTTTT
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
There are no transcripts associated with this gene.

Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002524.4 1154 UTR 3 NP_002515.1

View Full Product Details