Product Details
- SNP ID
-
rs143135847
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:231364624 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTACACAATTTTTTTCCATGTTTT[A/C]CATGAATAGCAATGGCAATCTTTTA
- Phenotype
-
MIM: 606425
MIM: 616086
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EGLN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs541569859] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EGLN1
- Gene Name
- egl-9 family hypoxia inducible factor 1
- Gene
- SPRTN
- Gene Name
- SprT-like N-terminal domain
There are no transcripts associated with this gene.
View Full Product Details