Product Details

SNP ID
rs143352734
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:78628013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTCTCTATAAGTCTAGTGTTC[A/C]TGGAGGTAGCATTGAAGATATGGTT
Phenotype
MIM: 613975
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFI44L PubMed Links

Gene Details

Gene
IFI44L
Gene Name
interferon induced protein 44 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006820.3 136 Missense Mutation CAT,CCT H33P NP_006811.2
XM_005270391.3 136 Intron XP_005270448.1
XM_005270392.4 136 Intron XP_005270449.1
XM_005270393.4 136 Intron XP_005270450.1
XM_006710304.3 136 Intron XP_006710367.1
XM_011540539.2 136 Missense Mutation CAT,CCT H33P XP_011538841.1
XM_017000120.1 136 Intron XP_016855609.1

View Full Product Details