Product Details
- SNP ID
-
rs143459066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:179368892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGAGTTCATTCCCAAAGAAGTT[C/T]TTCTTTCTCTGACCTATGCGGCCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AXDND1
PubMed Links
Gene Details
- Gene
- AXDND1
- Gene Name
- axonemal dynein light chain domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144696.5 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
NP_653297.3 |
XM_011509165.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507467.1 |
XM_011509166.2 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507468.1 |
XM_011509167.2 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507469.1 |
XM_011509168.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507470.1 |
XM_011509169.1 |
556 |
Missense Mutation |
CTT,TTT |
L43F |
XP_011507471.1 |
XM_011509170.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507472.1 |
XM_011509171.1 |
556 |
Missense Mutation |
CTT,TTT |
L22F |
XP_011507473.1 |
XM_011509174.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507476.1 |
XM_011509175.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507477.1 |
XM_011509176.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507478.1 |
XM_011509178.2 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507480.1 |
XM_011509179.2 |
556 |
Intron |
|
|
XP_011507481.1 |
XM_011509180.1 |
556 |
Missense Mutation |
CTT,TTT |
L64F |
XP_011507482.1 |
XM_011509181.2 |
556 |
Intron |
|
|
XP_011507483.1 |
XM_017000257.1 |
556 |
Intron |
|
|
XP_016855746.1 |
XM_017000258.1 |
556 |
Intron |
|
|
XP_016855747.1 |
XM_017000259.1 |
556 |
Intron |
|
|
XP_016855748.1 |
View Full Product Details