Product Details

SNP ID
rs143471064
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:44654733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGTCTCTGGCCAGGACTACTTC[A/G]TCAGCCCTCGAGTAGAGGTAGAGCT
Phenotype
MIM: 616136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF220 PubMed Links

Gene Details

Gene
RNF220
Gene Name
ring finger protein 220
There are no transcripts associated with this gene.

Gene
TMEM53
Gene Name
transmembrane protein 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300746.1 1006 Silent Mutation GAC,GAT D147D NP_001287675.1
NM_001300747.1 1006 Silent Mutation GAC,GAT D190D NP_001287676.1
NM_001300748.1 1006 Silent Mutation GAC,GAT D189D NP_001287677.1
NM_024587.3 1006 Silent Mutation GAC,GAT D220D NP_078863.2
XM_011542138.2 1006 Silent Mutation GAC,GAT D103D XP_011540440.1

View Full Product Details