Product Details
- SNP ID
-
rs143813756
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:153775684 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCGCCGACCCGGAAGGTCCCGAG[G/T]GGGGCTGCAGCCTGGCCTGGCGCCT
- Phenotype
-
MIM: 611347
MIM: 604193
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
INTS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34527123] are located under a probe and SNP(s) [rs36064263] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INTS3
- Gene Name
- integrator complex subunit 3
- Gene
- SLC27A3
- Gene Name
- solute carrier family 27 member 3
View Full Product Details