Product Details

SNP ID
rs143863599
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:151038871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGTCTTTCTGCCCCAGAGTTGC[A/G]GCTGAGTCTGACTAAGGGGCCTGGA
Phenotype
MIM: 611275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159642.1 703 Missense Mutation CAG,CGG Q11R NP_001153114.1
NM_138278.3 703 Missense Mutation CAG,CGG Q93R NP_612122.2
XM_011509236.1 703 Missense Mutation CAG,CGG Q93R XP_011507538.1
XM_017000420.1 703 Intron XP_016855909.1
Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
There are no transcripts associated with this gene.

Gene
PRUNE1
Gene Name
prune exopolyphosphatase
There are no transcripts associated with this gene.

View Full Product Details