Product Details

SNP ID
rs143873128
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:209830956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGGAAGAAGACAGTATTGTA[C/G]ATGATGCAGAAATGAACGATGAAGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIEXF PubMed Links

Gene Details

Gene
DIEXF
Gene Name
digestive organ expansion factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014388.6 398 Missense Mutation CAT,GAT H101D NP_055203.4
XM_006711275.3 398 Missense Mutation CAT,GAT H101D XP_006711338.1
XM_011509418.2 398 UTR 5 XP_011507720.1

View Full Product Details