Product Details

SNP ID
rs143975777
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:30939873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTAAAAAAATAAAAACTTCTT[C/T]TTTTTTAAATTGTTTTTGAATTTGT
Phenotype
MIM: 607204
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PUM1 PubMed Links

Gene Details

Gene
PUM1
Gene Name
pumilio RNA binding family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001020658.1 Intron NP_001018494.1
NM_014676.2 Intron NP_055491.1
Gene
SNORD103A
Gene Name
small nucleolar RNA, C/D box 103A
There are no transcripts associated with this gene.

Gene
SNORD103B
Gene Name
small nucleolar RNA, C/D box 103B
There are no transcripts associated with this gene.

View Full Product Details