Product Details

SNP ID
rs144076109
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:223394273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTGTGGCCCTGGTGCTGACCCG[A/T]CTCAAGAAGGCCCAGAGGATACGGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC185 PubMed Links

Gene Details

Gene
CCDC185
Gene Name
coiled-coil domain containing 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152610.2 901 Silent Mutation CGA,CGT R266R NP_689823.2

View Full Product Details