Product Details

SNP ID
rs144088253
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15660008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATCAAGCCCAGTGACTCAGATC[A/G]CATTGAACCTAAAGCTGTGAAGGCT
Phenotype
MIM: 601966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDI2 PubMed Links

Gene Details

Gene
DDI2
Gene Name
DNA damage inducible 1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032341.4 296 UTR 3 NP_115717.3
Gene
RSC1A1
Gene Name
regulatory solute carrier protein, family 1, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006511.2 296 Missense Mutation CAC,CGC H47R NP_006502.1

View Full Product Details