Product Details

SNP ID
rs144231687
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:214614999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAACTGAATTCAGGCAAAAAACA[A/G]ATAGAAAAACTGGAACAGGAACTTA
Phenotype
MIM: 600236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPF PubMed Links

Gene Details

Gene
CENPF
Gene Name
centromere protein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016343.3 491 Silent Mutation CAA,CAG Q110Q NP_057427.3
XM_011509082.2 491 Silent Mutation CAA,CAG Q110Q XP_011507384.1
XM_017000086.1 491 Silent Mutation CAA,CAG Q110Q XP_016855575.1

View Full Product Details