Product Details

SNP ID
rs144830172
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:206685445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCACCAGCCAGGTCCTGGGGCT[C/G]GGCATCAACGGCAAAGTTTTGCAGA
Phenotype
MIM: 602006
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MAPKAPK2 PubMed Links

Gene Details

Gene
MAPKAPK2
Gene Name
mitogen-activated protein kinase-activated protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004759.4 426 Silent Mutation CTC,CTG L72L NP_004750.1
NM_032960.3 426 Silent Mutation CTC,CTG L72L NP_116584.2
XM_005273353.3 426 Silent Mutation CTC,CTG L72L XP_005273410.1
XM_017002810.1 426 Intron XP_016858299.1

View Full Product Details