Product Details

SNP ID
rs144894067
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:112703107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCTGCCCTGCTGTGTCCCACA[A/G]AGCCAGCTCCACCTGACACACAGGG
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 502 Intron NP_001123551.1
NM_001286072.1 502 Intron NP_001273001.1
NM_001321324.1 502 Intron NP_001308253.1
NM_020963.4 502 Intron NP_066014.1
XM_005270869.4 502 Intron XP_005270926.1
XM_005270870.4 502 Intron XP_005270927.1
XM_017001318.1 502 Intron XP_016856807.1
XM_017001319.1 502 Intron XP_016856808.1
Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 502 Silent Mutation CTG,TTG L57L NP_001036143.1
NM_001042679.1 502 Silent Mutation CTG,TTG L57L NP_001036144.1
NM_175744.4 502 Silent Mutation CTG,TTG L57L NP_786886.1

View Full Product Details