Product Details

SNP ID
rs144977463
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:205569126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGCCGCAACCTGCAGCCCACGCT[C/T]ACCTACTGGAGCGTCTTCTTCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD4A PubMed Links

Gene Details

Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 143 Silent Mutation CTC,CTT L19L NP_857595.3
XM_011509229.2 143 Silent Mutation CTC,CTT L19L XP_011507531.1

View Full Product Details