Product Details

SNP ID
rs145116599
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:40626583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACAGGCGCCTGGTGAGGGATCC[A/G]AGTGTGGAGTCTGCCACTGAGGAGG
Phenotype
MIM: 611600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIMS3 PubMed Links

Gene Details

Gene
RIMS3
Gene Name
regulating synaptic membrane exocytosis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014747.2 968 Silent Mutation CTC,CTT L287L NP_055562.2
XM_011542479.2 968 Silent Mutation CTC,CTT L291L XP_011540781.1

View Full Product Details