Product Details

SNP ID
rs145207343
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:248349052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACAGCACCACCATTTCTAAGG[C/T]GGGATGTGTAGCTCAGGTCTTCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14C36 PubMed Links

Gene Details

Gene
OR14C36
Gene Name
olfactory receptor family 14 subfamily C member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001918.1 278 Missense Mutation GCG,GTG A93V NP_001001918.1

View Full Product Details