Product Details

SNP ID
rs145285600
Assay Type
Functionally Tested
NCBI dbSNP Submissions
10
Location
Chr.1:34761508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCCTACAGCTCATCCTGGTCA[C/T]GTGCCCCTCACTGCTCGTGGTCATG
Phenotype
MIM: 605425 MIM: 604493
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GJB4 PubMed Links
Additional Information
For this assay, SNP(s) [rs76188300] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GJB4
Gene Name
gap junction protein beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153212.2 912 Missense Mutation ACG,ATG T85M NP_694944.1
XM_011540679.2 912 Missense Mutation ACG,ATG T85M XP_011538981.1
Gene
GJB5
Gene Name
gap junction protein beta 5
There are no transcripts associated with this gene.

View Full Product Details