Product Details

SNP ID
rs145354886
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:179592654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGTATACAGGAATGTCTGCGGAA[A/G]GAAATAAGGTCACTTCTCATTTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD5 PubMed Links

Gene Details

Gene
TDRD5
Gene Name
tudor domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199085.1 557 Silent Mutation AAA,AAG K13K NP_001186014.1
NM_001199089.1 557 Silent Mutation AAA,AAG K13K NP_001186018.1
NM_001199091.1 557 Silent Mutation AAA,AAG K13K NP_001186020.1
NM_001199092.1 557 Intron NP_001186021.1
NM_173533.3 557 Silent Mutation AAA,AAG K13K NP_775804.2
XM_005244934.1 557 Silent Mutation AAA,AAG K13K XP_005244991.1
XM_005244935.4 557 Silent Mutation AAA,AAG K13K XP_005244992.1
XM_017000473.1 557 Silent Mutation AAA,AAG K13K XP_016855962.1

View Full Product Details