Product Details

SNP ID
rs145371506
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:3890748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGGATTCTTCTGCTCCGGACC[C/T]GGCACTGTGTTTTGGCACGGACAGG
Phenotype
MIM: 601883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf174 PubMed Links

Gene Details

Gene
C1orf174
Gene Name
chromosome 1 open reading frame 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207356.2 3034 Missense Mutation AGG,GGG R147G NP_997239.2
XM_005244743.3 3034 Missense Mutation AGG,GGG R158G XP_005244800.1
XM_011541323.2 3034 Missense Mutation AGG,GGG R158G XP_011539625.1
Gene
DFFB
Gene Name
DNA fragmentation factor subunit beta
There are no transcripts associated with this gene.

Gene
LINC01134
Gene Name
long intergenic non-protein coding RNA 1134
There are no transcripts associated with this gene.

View Full Product Details