Product Details

SNP ID
rs145462928
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:154963842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGGTTGCTGTAGACACAGTTC[A/G]CTGCCCGCAGAGATGATGGGCAAGT
Phenotype
MIM: 606903 MIM: 600560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928120 PubMed Links

Gene Details

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
There are no transcripts associated with this gene.

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
There are no transcripts associated with this gene.

Gene
PYGO2
Gene Name
pygopus family PHD finger 2
There are no transcripts associated with this gene.

Gene
SHC1
Gene Name
SHC adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130040.1 3127 Intron NP_001123512.1
NM_001130041.1 3127 Intron NP_001123513.1
NM_001202859.1 3127 Intron NP_001189788.1
NM_003029.4 3127 Intron NP_003020.2
NM_183001.4 3127 Intron NP_892113.4
XM_005245449.4 3127 Intron XP_005245506.1
XM_005245451.4 3127 Intron XP_005245508.1
XM_011509892.2 3127 Silent Mutation AGC,AGT S581S XP_011508194.1
XM_011509893.2 3127 Silent Mutation AGC,AGT S580S XP_011508195.1
XM_011509894.2 3127 Silent Mutation AGC,AGT S563S XP_011508196.1
XM_011509897.1 3127 Intron XP_011508199.1
XM_011509898.2 3127 Intron XP_011508200.1
XM_017002081.1 3127 Silent Mutation AGC,AGT S554S XP_016857570.1
XM_017002082.1 3127 Silent Mutation AGC,AGT S553S XP_016857571.1
XM_017002083.1 3127 Intron XP_016857572.1

View Full Product Details