Product Details

SNP ID
rs145641137
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:36140115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTTCATCATTTTCATAGTCCT[G/T]ACATAGCTGGGTGACCAGGGCACCA
Phenotype
MIM: 610955
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAPPC3 PubMed Links

Gene Details

Gene
TRAPPC3
Gene Name
trafficking protein particle complex 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270894.1 230 Missense Mutation AAG,CAG K32Q NP_001257823.1
NM_001270895.1 230 UTR 5 NP_001257824.1
NM_001270896.1 230 UTR 5 NP_001257825.1
NM_001270897.1 230 Intron NP_001257826.1
NM_014408.4 230 Missense Mutation AAG,CAG K32Q NP_055223.1

View Full Product Details