Product Details

SNP ID
rs145644780
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:248812228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACGGGGGAGGAGCGCCGAGGG[C/T]CCAGGGGGTGGGGAGGCAGACCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3BP5L PubMed Links

Gene Details

Gene
SH3BP5L
Gene Name
SH3 binding domain protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322462.1 1294 Missense Mutation GAC,GGC D166G NP_001309391.1
NM_001322463.1 1294 Missense Mutation GAC,GGC D166G NP_001309392.1
NM_001322464.1 1294 Missense Mutation GAC,GGC D118G NP_001309393.1
NM_030645.1 1294 Missense Mutation GAC,GGC D285G NP_085148.1
XM_017002404.1 1294 Missense Mutation GAC,GGC D143G XP_016857893.1
XM_017002405.1 1294 Missense Mutation GAC,GGC D143G XP_016857894.1
XM_017002406.1 1294 Missense Mutation GAC,GGC D143G XP_016857895.1
XM_017002407.1 1294 Missense Mutation GAC,GGC D253G XP_016857896.1

View Full Product Details