Product Details

SNP ID
rs145716190
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:86425422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAATATAAAGATTTTAATACC[A/T]GCCACATGGAAAGCTAATAATAACA
Phenotype
MIM: 604003
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 432 Silent Mutation CCA,CCT P90P NP_006527.1
XM_011542448.2 432 Silent Mutation CCA,CCT P90P XP_011540750.1

View Full Product Details