Product Details

SNP ID
rs145848446
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:151403296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGGCCCATCACATCTTGAGC[A/G]TTCCCCAGTCTGTGAAAGCCTATTA
Phenotype
MIM: 614787 MIM: 602177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGZ PubMed Links

Gene Details

Gene
POGZ
Gene Name
pogo transposable element with ZNF domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194937.1 6027 UTR 3 NP_001181866.1
NM_001194938.1 6027 UTR 3 NP_001181867.1
NM_015100.3 6027 UTR 3 NP_055915.2
NM_145796.3 6027 UTR 3 NP_665739.3
NM_207171.2 6027 UTR 3 NP_997054.1
XM_005244999.2 6027 Intron XP_005245056.1
XM_005245000.4 6027 Intron XP_005245057.1
XM_005245001.2 6027 Intron XP_005245058.1
XM_005245005.1 6027 UTR 3 XP_005245062.1
XM_005245006.4 6027 UTR 3 XP_005245063.1
XM_011509331.2 6027 Intron XP_011507633.1
XM_017000744.1 6027 UTR 3 XP_016856233.1
XM_017000745.1 6027 Intron XP_016856234.1
XM_017000746.1 6027 Intron XP_016856235.1
XM_017000747.1 6027 UTR 3 XP_016856236.1
XM_017000748.1 6027 UTR 3 XP_016856237.1
XM_017000749.1 6027 UTR 3 XP_016856238.1
Gene
PSMB4
Gene Name
proteasome subunit beta 4
There are no transcripts associated with this gene.

View Full Product Details