Product Details

SNP ID
rs146201991
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:160831413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGAGCAGGGTTCTGGGCTTTAGG[C/T]TGACTCTTTCCAATCTGCAAAAGAA
Phenotype
MIM: 605554 MIM: 600684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD244 PubMed Links

Gene Details

Gene
CD244
Gene Name
CD244 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166663.1 1272 Silent Mutation CAA,CAG Q349Q NP_001160135.1
NM_001166664.1 1272 Silent Mutation CAA,CAG Q252Q NP_001160136.1
NM_016382.3 1272 Silent Mutation CAA,CAG Q344Q NP_057466.1
XM_011509621.2 1272 Intron XP_011507923.1
XM_011509622.2 1272 Intron XP_011507924.1
XM_011509623.2 1272 Silent Mutation CAA,CAG Q146Q XP_011507925.1
Gene
LY9
Gene Name
lymphocyte antigen 9
There are no transcripts associated with this gene.

View Full Product Details