Product Details

SNP ID
rs146340487
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:152759616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGACCAGCAGCAGATCCAGTGC[C/T]GCCTGCCGCTCCAACAGTGCTGCGT
Phenotype
MIM: 613260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KPRP PubMed Links

Gene Details

Gene
KPRP
Gene Name
keratinocyte proline rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025231.1 127 Missense Mutation CGC,TGC R10C NP_001020402.1
XM_011509571.1 127 Missense Mutation CGC,TGC R10C XP_011507873.1

View Full Product Details