Product Details
- SNP ID
-
rs146841294
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:77783325 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGCTAAGAGTGTTTGATCTTCCT[C/T]TGACTTGGTACTATTCTCTCTCCCA
- Phenotype
-
MIM: 616773
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIGA1
PubMed Links
Gene Details
- Gene
- MIGA1
- Gene Name
- mitoguardin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270384.1 |
171 |
Silent Mutation |
CTG,TTG |
L57L |
NP_001257313.1 |
NM_198549.3 |
171 |
Silent Mutation |
CTG,TTG |
L57L |
NP_940951.1 |
XM_005270846.4 |
171 |
Silent Mutation |
CTG,TTG |
L57L |
XP_005270903.1 |
XM_005270847.4 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_005270904.1 |
XM_005270848.4 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_005270905.1 |
XM_011541422.2 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_011539724.1 |
XM_017001227.1 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_016856716.1 |
XM_017001228.1 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_016856717.1 |
XM_017001229.1 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_016856718.1 |
XM_017001230.1 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_016856719.1 |
XM_017001231.1 |
171 |
Silent Mutation |
CTG,TTG |
L57L |
XP_016856720.1 |
XM_017001232.1 |
171 |
Silent Mutation |
CTG,TTG |
L25L |
XP_016856721.1 |
View Full Product Details