Product Details

SNP ID
rs146956806
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:229432878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGACCCATACCGACCATGAC[A/G]CCCTGCAGAGCCGAGACACCACGCA
Phenotype
MIM: 102610 MIM: 607613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTA1 PubMed Links

Gene Details

Gene
ACTA1
Gene Name
actin, alpha 1, skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100.3 237 Silent Mutation GGC,GGT G44G NP_001091.1
Gene
NUP133
Gene Name
nucleoporin 133
There are no transcripts associated with this gene.

View Full Product Details