Product Details

SNP ID
rs146969788
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:16232234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCACCCGTAGCAGGGGCAGCTC[C/T]CAGGCCTGCCGGAAGGCTGTGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO7P1 PubMed Links

Gene Details

Gene
ANO7P1
Gene Name
anoctamin 7 pseudogene 1
There are no transcripts associated with this gene.

Gene
RSG1
Gene Name
REM2 and RAB like small GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030907.3 1013 Nonsense Mutation TGA,TGG *197W NP_112169.2
XM_011542126.2 1013 Missense Mutation GAG,GGG E201G XP_011540428.1

View Full Product Details