Product Details

SNP ID
rs147089521
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:37566979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAGCGCACACCAACTTTTTTC[A/G]GCCGTTGCTGTCGTACTGCTCGCCT
Phenotype
MIM: 602135 MIM: 609365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 2235 Intron NP_003453.2
XM_005271172.2 2235 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323623.1 2235 Missense Mutation CCG,CTG P759L NP_001310552.1
NM_001323624.1 2235 Missense Mutation CCG,CTG P508L NP_001310553.1
NM_013285.2 2235 Missense Mutation CCG,CTG P691L NP_037417.1

View Full Product Details