Product Details

SNP ID
rs147244512
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:155039509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGGCTCTTTGTCCTGGGATA[C/T]GCCTTGGCTGCCATCTATGTGGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCST1 PubMed Links

Gene Details

Gene
DCST1
Gene Name
DC-STAMP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143687.2 483 Silent Mutation TAC,TAT Y98Y NP_001137159.1
NM_152494.3 483 Silent Mutation TAC,TAT Y123Y NP_689707.2
Gene
DCST2
Gene Name
DC-STAMP domain containing 2
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details