Product Details

SNP ID
rs147368333
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:1918353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATGTGGCTGGGCCTGTGCCTG[C/G]TTCCGGAGGCAGCAGAACCGGACAC
Phenotype
MIM: 610171
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
There are no transcripts associated with this gene.

Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
There are no transcripts associated with this gene.

Gene
TMEM52
Gene Name
transmembrane protein 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178545.3 285 Silent Mutation AAC,AAG N83K NP_848640.1

View Full Product Details