Product Details

SNP ID
rs147417577
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:110339876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTCACGTTCCTCTGGGGCCGC[C/T]TCCTCAGCTCCCGGCGGCGGGGACG
Phenotype
MIM: 606077
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC440600 PubMed Links

Gene Details

Gene
LOC440600
Gene Name
uncharacterized LOC440600
There are no transcripts associated with this gene.

Gene
RBM15
Gene Name
RNA binding motif protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201545.1 554 Silent Mutation GCC,GCT A157A NP_001188474.1
NM_022768.4 554 Silent Mutation GCC,GCT A157A NP_073605.4

View Full Product Details